Learn More
eMolecules​ 1257849-07-2 | (2S)-3-(5-chloro-1H-indol-3-yl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid | Synthonix | MFCD16883066 | 460.910 | C26H21ClN2O4 | 97.000 | OC(=O)[C@H](Cc1c[nH]c2ccc(Cl)cc12)NC(=O)OCC1c2ccccc2-c2ccccc12 | 5g | 831794428
Catalog No. 5000158919 Shop All eMolecules Inc. Products
Encompass_Preferred
(2S)-3-(5-chloro-1H-indol-3-yl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid | Synthonix | 1257849-07-2 | MFCD16883066 | 460.910 | C26H21ClN2O4 | 97.000 | OC(=O)[C@H](Cc1c[nH]c2ccc(Cl)cc12)NC(=O)OCC1c2ccccc2-c2ccccc12 | 5g | 831794428
Catalog No. 50-001-58919 Supplier eMolecules​ Supplier No. 831794428
(2S)-3-(5-chloro-1H-indol-3-yl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid | Synthonix | 1257849-07-2 | MFCD16883066 | 460.910 | C26H21ClN2O4 | 97.000 | OC(=O)[C@H](Cc1c[nH]c2ccc(Cl)cc12)NC(=O)OCC1c2ccccc2-c2ccccc12 | 5g | 831794428