Learn More
Sino Biological Recombinant Human FABP5 Protein
Supplier:  Sino Biological 12581HNAE100
Encompass_Preferred
A DNA sequence encoding the human FABP5 (Q01469) (Met 1-Glu 135) was expressed and purified.
Catalog No. 50-161-8069