Learn More
Sino Biological Recombinant Human C1qR1/CD93 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12589H08H50
Encompass_Preferred
A DNA sequence encoding the human CD93 (Q9NPY3) extracellular domain (Met 1-Lys 580) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8078