Learn More
Waters Corp SEC Guard Cartridge, 125Å, 2.5 μm, 4.6 mm × 30 mm
Supplier: Waters Corp 186011356
This SEC Guard Cartridge with MaxPeak Premier Technology is designed for biotherapeutic peptides and small proteins (1K to 80K Dalton). It offers higher resolution of size variants, reproducible HMWP quantitation, reduced run times, and solvent cost savings. This guard cartridge uses MaxPeak High Performance Surface (HPS) Technologies to minimize secondary ionic interactions between biomolecules and column hardware surfaces.
- Chemistry: SEC
- Separation mode: SEC
- Particle substrate: Hybrid
- pH range: 2.5 to 8 pH
- Temperature limits: 60°C
- Maximum pressure: 11,000 psi (758 Bar)
- Particle size: 2.5 μm
- Pore size: 200 Å
- Molecular weight range: 1,000 to 80,000 Dalton
- Format: Guard column
- Dimensions: 4.6 mm × 30 mm
- System compatibility: UHPLC, UPLC, HPLC
- Application: Protein
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.