Learn More
Bellco Glass, Inc. 12.5cm2 Tissue Culture Flask
SDP
Catalog No. 502330509
Encompass_Preferred
DescriptionTissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachmentConvenient zip-closure bag is a practical alternative to folding and taping open bagsLarge opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapersStable base enables filling with flask standing uprightThick, durable wallsFlask is printed with lot number and expiration date for easy identification and trackingVent caps feature 0.22µm hydrophobic membrane Pack Description10/Re-sealable BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorFlask: ClearSecondary ColorCap: GreenPrimary MaterialFlask: PolystyreneSecondary MaterialCap: HDPEMaximum Volume25mLWorking Volume2-4mLGraduatedYesTissue Culture TreatedYesCell Growth Area12.5cm2RNase/DNase FreeYes
Catalog No. 50-233-0509 Supplier Bellco Glass, Inc. Supplier No. 229321000
DescriptionTissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachmentConvenient zip-closure bag is a practical alternative to folding and taping open bagsLarge opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapersStable base enables filling with flask standing uprightThick, durable wallsFlask is printed with lot number and expiration date for easy identification and trackingVent caps feature 0.22µm hydrophobic membrane Pack Description10/Re-sealable BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorFlask: ClearSecondary ColorCap: GreenPrimary MaterialFlask: PolystyreneSecondary MaterialCap: HDPEMaximum Volume25mLWorking Volume2-4mLGraduatedYesTissue Culture TreatedYesCell Growth Area12.5cm2RNase/DNase FreeYes