Learn More
Ace Glass, Inc. 125mL 15mm equalizing add funnel
SDP
Catalog No. 50974783
Encompass_Preferred
125mL, 15mm equalizing addition funnel with graduations for use with ACE pressure reactors. Funnel is heavy walled and features a #15 Ace-Thred at the top, bottom drip tip, pressure equalizing arm, and 0-3mm threaded PTFE stopcock plug.
Catalog No. 50-974-783 Supplier Ace Glass, Inc. Supplier No. 729912
125mL, 15mm equalizing addition funnel with graduations for use with ACE pressure reactors. Funnel is heavy walled and features a #15 Ace-Thred at the top, bottom drip tip, pressure equalizing arm, and 0-3mm threaded PTFE stopcock plug.