Learn More
Ace Glass, Inc. 125mL 24/25 separatory funnel with polyethylene stopper
SDP
Catalog No. 50974690
Encompass_Preferred
125ml separatory funnel with polyethylene stopper. Outer ground joint at top is a 24/25 Rodaviss joint. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-690 Supplier Ace Glass, Inc. Supplier No. 723109
Add to Cart
Add to Cart
125ml separatory funnel with polyethylene stopper. Outer ground joint at top is a 24/25 Rodaviss joint. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.