Learn More
Ace Glass, Inc. 125mL 24/40 separatory funnel body only
SDP
Catalog No. 50974694
Encompass_Preferred
125ml separatory funnel with 24/40 outer ground Rodaviss joint at top. Funnel body supplied with stopcock plug.
Catalog No. 50-974-694 Supplier Ace Glass, Inc. Supplier No. 723136
125ml separatory funnel with 24/40 outer ground Rodaviss joint at top. Funnel body supplied with stopcock plug.