Learn More
Ace Glass, Inc. 125mL 34/45 extraction apparatus
SDP
Catalog No. 50973399
Encompass_Preferred
125ml Soxhlet extraction apparatus with 34/45 joint, uses Friedrichs Condenser in place of Allihn Condenser. Apparatus consists of flask cap, condenser, extractor, and flask.
Catalog No. 50-973-399 Supplier Ace Glass, Inc. Supplier No. 671815
125ml Soxhlet extraction apparatus with 34/45 joint, uses Friedrichs Condenser in place of Allihn Condenser. Apparatus consists of flask cap, condenser, extractor, and flask.