Learn More
Chemglass Life Sciences 125mL 4-Neck European Flask, 1-CN 24/40 Outer, 2-SN 14/20 Outer Angled 20°, 1-SN #7 Chem-Thread
SDP
Catalog No. CG157202
Encompass_Preferred
125mL 4-Neck European Flask, 1-CN 24/40 Outer, 2-SN 14/20 Outer Angled 20¿, 1-SN #7 Chem-Thread. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. With standard taper outer joints on center and side necks and a #7 Chem-Thread for insertion of thermometers or bleed tubes having an O.D. between 4 and 7mm.
Catalog No. CG157202 Supplier Chemglass Life Sciences Supplier No. CG157202
125mL 4-Neck European Flask, 1-CN 24/40 Outer, 2-SN 14/20 Outer Angled 20¿, 1-SN #7 Chem-Thread. European style flask with tapered walls and a shallow hemispherical bottom for more efficient mechanical or magnetic stirring. With standard taper outer joints on center and side necks and a #7 Chem-Thread for insertion of thermometers or bleed tubes having an O.D. between 4 and 7mm.