Learn More
Chemglass Life Sciences 125mL Addition Funnel, Graduated, 29/26 Joint Size, 2mm PTFE Metering Stpk, 295mm OAH
SDP
Catalog No. CG171314
Encompass_Preferred
125mL Addition Funnel, Graduated, 29/26 Joint Size, 2mm PTFE Metering Stopcock, 295mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.
Catalog No. CG171314 Supplier Chemglass Life Sciences Supplier No. CG171314
125mL Addition Funnel, Graduated, 29/26 Joint Size, 2mm PTFE Metering Stopcock, 295mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.