Learn More
Chemglass Life Sciences 125mL Addition Funnel, 24/40 Joint Size, 2mm Stpk, 325mm OAH
SDP
Catalog No. 50873262
Encompass_Preferred
125mL Addition Funnel, 24/40 Joint Size, 2mm Stopcock, 325mm OAH. Cylindrical shape addition funnel with PTFE stopcocks. For replacement stoppers see CG-3000.
Catalog No. 50-873-262 Supplier Chemglass Life Sciences Supplier No. CG170802
125mL Addition Funnel, 24/40 Joint Size, 2mm Stopcock, 325mm OAH. Cylindrical shape addition funnel with PTFE stopcocks. For replacement stoppers see CG-3000.