Learn More
Chemglass Life Sciences 125mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk
SDP
Catalog No. AF055103
Encompass_Preferred
125mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk. Unique design of this addition funnel permits solutions to be prepared using a magnetic Stir Bar. Side arm has a 2mm high vacuum glass T-Bore stopcock having a Stainless Steel constant pressure retention device for evacuation and purging of both the funnel and lower vessel. Lower valve is a 0-4mm Chem-Vac™ Chem-Cap valve. The top outer and lower inner standard taper joints are the same size. The 50mL and 125mL sizes are graduated in 1.0mL subdivisions, while the 250mL has 5.0mL subdivisions.
Catalog No. AF055103 Supplier Chemglass Life Sciences Supplier No. AF055103
125mL Funnel, Addition, Graduated, 24/40 Joint Size, Airfree, Schlenk. Unique design of this addition funnel permits solutions to be prepared using a magnetic Stir Bar. Side arm has a 2mm high vacuum glass T-Bore stopcock having a Stainless Steel constant pressure retention device for evacuation and purging of both the funnel and lower vessel. Lower valve is a 0-4mm Chem-Vac™ Chem-Cap valve. The top outer and lower inner standard taper joints are the same size. The 50mL and 125mL sizes are graduated in 1.0mL subdivisions, while the 250mL has 5.0mL subdivisions.