Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Ammonia N Calibration Standard, 1000 mg/L, 125 mL, Aqueous

Catalog No. 50465865 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Ammonia as N Ion Calibration Standard contains 1000 mg/L of Ammonia as N in H2O. It can be used for primary calibration or to prepare second source calibration check standards, and is traceable to NIST Standard Reference Materials where available. This CRM is covered by ISO 17034 accreditation and is guaranteed stable for one year.

  • Certified Reference Material (CRM).
  • Accredited to ISO 17034.
  • Suitable for EPA 350.1, EPA 350.2, EPA 350.3, SM 4500-NH3 B, SM 4500-NH3 C, SM 4500-NH3 D, SM 4500-NH3 E, SM 4500-NH3 F, SM 4500-NH3 G methods.
  • Storage at 20°C to 25°C (ambient).
  • Shelf life of 24 months from date of manufacture.
  • Minimum shelf life of 12 months on ship date.
  • One component per package.
  • Provided in a 125 mL poly bottle.
  • Liquid formulation.
  • Ideal for environmental applications.

Catalog No. 50-465-865 Supplier Waters Corp Supplier No. 045
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Ammonia as N Ion Calibration Standard contains 1000 mg/L of Ammonia as N in H2O. It can be used for primary calibration or to prepare second source calibration check standards, and is traceable to NIST Standard Reference Materials where available. This CRM is covered by ISO 17034 accreditation and is guaranteed stable for one year.

  • Certified Reference Material (CRM).
  • Accredited to ISO 17034.
  • Suitable for EPA 350.1, EPA 350.2, EPA 350.3, SM 4500-NH3 B, SM 4500-NH3 C, SM 4500-NH3 D, SM 4500-NH3 E, SM 4500-NH3 F, SM 4500-NH3 G methods.
  • Storage at 20°C to 25°C (ambient).
  • Shelf life of 24 months from date of manufacture.
  • Minimum shelf life of 12 months on ship date.
  • One component per package.
  • Provided in a 125 mL poly bottle.
  • Liquid formulation.
  • Ideal for environmental applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.