Learn More
Waters Corp Waters Corporation Ammonia as NH3 Standard, 1000 mg/L, 125 mL
Catalog No. 50465864 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Ammonia as NH3 Ion Calibration Standard has a concentration of 1000 mg/L in an H2O matrix. It is suitable for primary calibration or for preparing second-source calibration check standards.
  • Traceable to NIST Standard Reference Materials (where available)
  • Guaranteed stable for one year
Catalog No. 50-465-864 Supplier Waters Corp Supplier No. 044
This Ammonia as NH3 Ion Calibration Standard has a concentration of 1000 mg/L in an H2O matrix. It is suitable for primary calibration or for preparing second-source calibration check standards.
  • Traceable to NIST Standard Reference Materials (where available)
  • Guaranteed stable for one year