Learn More
Waters Corp 125 mL Ammonium N
Supplier:  Waters Corp 78104
Encompass_Preferred
The 125 mL Ammonium N Standard is intended to be used for cation analysis by Ion Chromatography. The standard can be used for primary calibration or to prepare second source calibration check standards. It is traceable to NIST Standard Reference Materials, where available, and guaranteed stable for one year. The 125 ml Ammonium N, Ion Chromatography Standard is 100 mg/L in H2O.The standard is provided in one 125 mL screw-cap bottle. Store at room temperature.
Catalog No. 50-465-884