Learn More
Waters Corp Waters Corporation Cation Calibration Standard, Ammonium as NH4, 100 mg/L, 125 mL
Catalog No. 50465883 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed for cation analysis by Ion Chromatography, this standard can be used for primary calibration or to prepare second source calibration check standards. It is traceable to NIST Standard Reference Materials, where available, and guaranteed stable for one year, for direct use with ion-exchange chromatography.
  • Ammonium as NH4 cation calibration standard 100 mg/L in H2O matrix.
  • Used for cation analysis by Ion Chromatography.
  • For primary calibration or to prepare second source calibration check standards.
  • Traceable to NIST Standard Reference Materials, where available.
  • Guaranteed stable for one year.
  • Designed for direct use with ion-exchange chromatography.
  • Helps align chromatograph for a linear relationship with the solution signal.
  • Offers 100 mg/L concentration.
  • Inspected and satisfies strict QC process.
Catalog No. 50-465-883 Supplier Waters Corp Supplier No. 78102
Designed for cation analysis by Ion Chromatography, this standard can be used for primary calibration or to prepare second source calibration check standards. It is traceable to NIST Standard Reference Materials, where available, and guaranteed stable for one year, for direct use with ion-exchange chromatography.
  • Ammonium as NH4 cation calibration standard 100 mg/L in H2O matrix.
  • Used for cation analysis by Ion Chromatography.
  • For primary calibration or to prepare second source calibration check standards.
  • Traceable to NIST Standard Reference Materials, where available.
  • Guaranteed stable for one year.
  • Designed for direct use with ion-exchange chromatography.
  • Helps align chromatograph for a linear relationship with the solution signal.
  • Offers 100 mg/L concentration.
  • Inspected and satisfies strict QC process.