Learn More
Ace Glass, Inc. 125mL complete gas washing bottle, bottle and stopper
SDP
Catalog No. 50978370
Encompass_Preferred
125mL Dreschel, high form, gas washing bottle with standard taper 24/40 joint. Bottle supplied with 24/40 stopper.
Catalog No. 50-978-370 Supplier Ace Glass, Inc. Supplier No. 551608
125mL Dreschel, high form, gas washing bottle with standard taper 24/40 joint. Bottle supplied with 24/40 stopper.