Learn More
Chemglass Life Sciences Bottle Only, 125mL Gas Washing
SDP
Catalog No. CG111205
Encompass_Preferred
Bottle Only, 125mL Gas Washing. Replacement part for item CG111201. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and plain tube for gas dispersion.
Catalog No. CG111205 Supplier Chemglass Life Sciences Supplier No. CG111205
Bottle Only, 125mL Gas Washing. Replacement part for item CG111201. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and plain tube for gas dispersion.