Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Ion Calibration Standard, Bromate, 1000 mg/L, 125 mL

Catalog No. 50465873 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This is a 125 mL liquid standard containing Bromate at a concentration of 1000 mg/L in H2O. It can be used for primary calibration or to prepare second source calibration check standards. This certified reference material is traceable to NIST Standard Reference Materials and is covered by ISO 17034 accreditation.

  • Liquid formulation in H2O, 125 mL volume
  • Bromate concentration: 1000 mg/L
  • For environmental applications and primary/secondary calibration
  • Traceable to NIST SRMs, guaranteed stable for one year
  • ISO 17034 accredited CRM
  • Supports methods EPA 300.0, EPA 300.1, SM 4110 B
  • Store at 4 ± 2°C
  • Shelf life: 24 months from manufacture (12 months min. on ship)
  • Hazardous material; special freight/charges may apply

Catalog No. 50-465-873 Supplier Waters Corp Supplier No. 065
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This is a 125 mL liquid standard containing Bromate at a concentration of 1000 mg/L in H2O. It can be used for primary calibration or to prepare second source calibration check standards. This certified reference material is traceable to NIST Standard Reference Materials and is covered by ISO 17034 accreditation.

  • Liquid formulation in H2O, 125 mL volume
  • Bromate concentration: 1000 mg/L
  • For environmental applications and primary/secondary calibration
  • Traceable to NIST SRMs, guaranteed stable for one year
  • ISO 17034 accredited CRM
  • Supports methods EPA 300.0, EPA 300.1, SM 4110 B
  • Store at 4 ± 2°C
  • Shelf life: 24 months from manufacture (12 months min. on ship)
  • Hazardous material; special freight/charges may apply

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.