Learn More
Waters Corp Waters Corporation Calcium Standard, 1000 mg/L, Unpreserved, 125 mL
Catalog No. 50466721 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Calcium Cation Calibration Standard is 1000 mg/L with an H2O matrix. It is designed for cation analysis using Ion Chromatography and can be used for primary calibration or to prepare second source calibration check standards. The standard is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.
  • Formulation: Liquid
  • Volume: 125 mL
  • Designed for cation analysis using Ion Chromatography
  • Can be used for primary calibration or to prepare second source calibration check standards
  • Traceable to NIST Standard Reference Materials, where available
  • Guaranteed stable for one year
  • Units per package: 1 pk
Catalog No. 50-466-721 Supplier Waters Corp Supplier No. K10
Add to Cart
Add to Cart
This Calcium Cation Calibration Standard is 1000 mg/L with an H2O matrix. It is designed for cation analysis using Ion Chromatography and can be used for primary calibration or to prepare second source calibration check standards. The standard is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.
  • Formulation: Liquid
  • Volume: 125 mL
  • Designed for cation analysis using Ion Chromatography
  • Can be used for primary calibration or to prepare second source calibration check standards
  • Traceable to NIST Standard Reference Materials, where available
  • Guaranteed stable for one year
  • Units per package: 1 pk