Learn More
Waters Corp WATERS CORPORATION Chloride Ion Calibration Standard, 1000 mg/L, 125 mL, H2O Matrix
Catalog No. 50465867 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Chloride Ion Calibration Standard 1000 mg/L with a matrix of H2O is designed for primary calibration or to prepare second source calibration check standards. This standard is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.
  • Can be used for primary calibration.
  • Can be used to prepare second source calibration check standards.
  • Traceable to NIST Standard Reference Materials (where available).
  • Guaranteed stable for one year.
Catalog No. 50-465-867 Supplier Waters Corp Supplier No. 047
The Chloride Ion Calibration Standard 1000 mg/L with a matrix of H2O is designed for primary calibration or to prepare second source calibration check standards. This standard is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.
  • Can be used for primary calibration.
  • Can be used to prepare second source calibration check standards.
  • Traceable to NIST Standard Reference Materials (where available).
  • Guaranteed stable for one year.