Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Ion Calibration Standard, Chlorite, 1000 mg/L, 125 mL

Catalog No. 50465875 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Chlorite Ion Calibration Standard, with a concentration of 1000 mg/L in an H2O matrix, is designed for primary calibration or for preparing second source calibration check standards. It is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.

  • Formulation: Liquid
  • Application: Environment
  • Traceability to NIST Standard Reference Materials, where available
  • Guaranteed stable for one year

Catalog No. 50-465-875 Supplier Waters Corp Supplier No. 067
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Chlorite Ion Calibration Standard, with a concentration of 1000 mg/L in an H2O matrix, is designed for primary calibration or for preparing second source calibration check standards. It is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.

  • Formulation: Liquid
  • Application: Environment
  • Traceability to NIST Standard Reference Materials, where available
  • Guaranteed stable for one year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.