Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Hexavalent Chromium Standard, 1000 mg/L, 125 mL, ISO 17034

Catalog No. 50465842 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product is a Hexavalent Chromium standard, designed for analytical applications. It is a Certified Reference Material (CRM) with ISO 17034 accreditation, providing a reliable solution for various environmental testing methods.

  • Contains Hexavalent Chromium at 1000 mg/L.
  • Provided in a 125 mL volume.
  • Aqueous (H2O) matrix.
  • Certified Reference Material (CRM).
  • Accredited to ISO 17034.
  • Suitable for methods including EPA 218.4, EPA 218.5, EPA 7195, EPA 7196A, EPA 7197, EPA 7198, EPA 7199, SM 3500-Cr D.
  • Storage at 20–25°C (Ambient).
  • Shelf life of 36 months from date of manufacture.
  • Minimum shelf life on ship date of 12 months.
  • Packaged in a 125 mL poly bottle.
  • Ideal for environmental applications.

Catalog No. 50-465-842 Supplier Waters Corp Supplier No. 019
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This product is a Hexavalent Chromium standard, designed for analytical applications. It is a Certified Reference Material (CRM) with ISO 17034 accreditation, providing a reliable solution for various environmental testing methods.

  • Contains Hexavalent Chromium at 1000 mg/L.
  • Provided in a 125 mL volume.
  • Aqueous (H2O) matrix.
  • Certified Reference Material (CRM).
  • Accredited to ISO 17034.
  • Suitable for methods including EPA 218.4, EPA 218.5, EPA 7195, EPA 7196A, EPA 7197, EPA 7198, EPA 7199, SM 3500-Cr D.
  • Storage at 20–25°C (Ambient).
  • Shelf life of 36 months from date of manufacture.
  • Minimum shelf life on ship date of 12 months.
  • Packaged in a 125 mL poly bottle.
  • Ideal for environmental applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.