Learn More
Waters Corp Waters QC Trace Metal Standard, Copper, 1000 mg/L, 125 mL
Catalog No. 50465845 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This copper QC trace metal standard is provided at a concentration of 1000 mg/L in an HNO3 matrix. It is suitable for various analytical techniques, offering reliability for laboratory applications.
  • QC trace metal standard
  • Use with ICP-OES, ICP-MS and AA methods
  • One 15 mL screw-cap vial yields up to 2 L after dilution
  • Formulated as liquid
  • Designed for environmental applications
  • Contains hazardous material which requires special freight handling
Catalog No. 50-465-845 Supplier Waters Corp Supplier No. 022
Add to Cart
Add to Cart
This copper QC trace metal standard is provided at a concentration of 1000 mg/L in an HNO3 matrix. It is suitable for various analytical techniques, offering reliability for laboratory applications.
  • QC trace metal standard
  • Use with ICP-OES, ICP-MS and AA methods
  • One 15 mL screw-cap vial yields up to 2 L after dilution
  • Formulated as liquid
  • Designed for environmental applications
  • Contains hazardous material which requires special freight handling