Learn More
Bellco Glass, Inc. 125mL Erlenmeyer Flask, Vent Cap, Plain Bottom, PETG, Sterile
SDP
Supplier:  Bellco Glass, Inc. 229801000
Encompass_Preferred
Designed for mammalian suspension culture applications as well as bacterial & yeast cultureAvailable with either solid or vent cap to eliminate the chance of unintended venting, unlike combination capsVent caps feature 0.22µm hydrophobic membrane which allows for gas exchange without contamination; over-sized vent area in cap improves gas exchangePackaged sterile in individual bags for easy storage and handlingManufactured from medical grade virgin PETG which reduces gas permeability Pack Description1/BagCase Quantity12SterileYesNon-pyrogenicYesAutoclavableNoPrimary ColorFlask: ClearSecondary ColorCap: GreenPrimary MaterialFlask: PETGSecondary MaterialCap: HDPEMaximum Volume125mLWorking Volume30mLGraduatedYesGraduation Increments25mLTissue Culture TreatedNoRNase/DNase FreeYes
Catalog No. 50-232-9468