Learn More
Chemglass Life Sciences 125mL Erlenmeyer Flask, 24-410 GPI Screw Thread
SDP
Catalog No. 501531164
Encompass_Preferred
125mL Erlenmeyer Flask, 24-410 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.
Catalog No. 50-153-1164 Supplier Chemglass Life Sciences Supplier No. CG154302
125mL Erlenmeyer Flask, 24-410 GPI Screw Thread. Erlenmeyer flask with screw thread finish. Supplied complete with white rubber lined black phenolic cap. Long skirted caps can be loosened without falling off while shaking flask.