Learn More
Ace Glass, Inc. 125mL graduated cylindrical addition funnel
SDP
Catalog No. 50979231
Encompass_Preferred
125mL graduated cylindrical addition funnel with a 24/40 standard taper stopper joint. Stopcock and Stopper not supplied.
Catalog No. 50-979-231 Supplier Ace Glass, Inc. Supplier No. 726250
125mL graduated cylindrical addition funnel with a 24/40 standard taper stopper joint. Stopcock and Stopper not supplied.