Learn More
Ace Glass, Inc. 125mL gas washing bottle without stopper
SDP
Catalog No. 50971890
Encompass_Preferred
125mL Dreschel, high form, gas washing bottle with standard taper 24/40 joint.
Catalog No. 50-971-890 Supplier Ace Glass, Inc. Supplier No. 551605
125mL Dreschel, high form, gas washing bottle with standard taper 24/40 joint.