Learn More
Chemglass Life Sciences Complete 125mL Bottle, Gas Washing, Tall Form, Drechel, Approx 290mm OAH
SDP
Catalog No. CG111201
Encompass_Preferred
Complete 125mL Bottle, Gas Washing, Tall Form, Drechel, Approx 290mm OAH. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and plain tube for gas dispersion. Stopper is held in place with two CG-110-02, 18mm stainless steel springs. Springs are supplied when bottle is ordered complete.
Catalog No. CG111201 Supplier Chemglass Life Sciences Supplier No. CG111201
Complete 125mL Bottle, Gas Washing, Tall Form, Drechel, Approx 290mm OAH. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and plain tube for gas dispersion. Stopper is held in place with two CG-110-02, 18mm stainless steel springs. Springs are supplied when bottle is ordered complete.