Learn More
Ace Glass, Inc. 125mL hemispherical fabric mantle, Glas-Col, 70mm max flask diameter, 1.63in inside depth, 4.25in OD, 2.38in height, 80 watts, CSA rated
SDP
Catalog No. 50977265
Encompass
125mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.
Catalog No. 50-977-265 Supplier Ace Glass, Inc. Supplier No. 1203509
125mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.