Learn More
Foxx Life Sciences 125mL Wide Mouth LDPE Reagent Bottles, 72CS
Catalog No. 5000846707
Encompass_Preferred
EZLabpure™  125mL LDPE Reagent Bottles Non Sterile provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Wide Mouth LDPE Reagent Bottle features large openings for easy filling, cleaning & rapid content transfer. LDPE chemical resistant reagent bottles offer excel chemical resistance and have high impact strength
Features
LDPE storage reagent bottles are flexible and squeezable for easy dispensing.
LDPE storage bottles are lightweight and crack-resistant.
LDPE Bottles with low Liquid Retention
Leak-proof Linerless screw caps for secure sealing.
Catalog No. 50-008-46707 Supplier Foxx Life Sciences Supplier No. 7642B11FLS
EZLabpure™  125mL LDPE Reagent Bottles Non Sterile provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Wide Mouth LDPE Reagent Bottle features large openings for easy filling, cleaning & rapid content transfer. LDPE chemical resistant reagent bottles offer excel chemical resistance and have high impact strength
Features
LDPE storage reagent bottles are flexible and squeezable for easy dispensing.
LDPE storage bottles are lightweight and crack-resistant.
LDPE Bottles with low Liquid Retention
Leak-proof Linerless screw caps for secure sealing.