Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Standard, Lead, 1000 mg/L, 125 mL

Catalog No. 50465847 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Lead, QC Trace Metal Standard 1000 mg/L Matrix HNO3. This standard is designed for use with ICP-OES, ICP-MS and AA methods. One 15 mL screw-cap vial yields up to 2 liters after dilution.

  • Formulation: Liquid
  • Application: Environment
  • Product Type: Standards
  • This product contains hazardous material which requires special freight handling. Additional charges may apply.

Catalog No. 50-465-847 Supplier Waters Corp Supplier No. 024
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Lead, QC Trace Metal Standard 1000 mg/L Matrix HNO3. This standard is designed for use with ICP-OES, ICP-MS and AA methods. One 15 mL screw-cap vial yields up to 2 liters after dilution.

  • Formulation: Liquid
  • Application: Environment
  • Product Type: Standards
  • This product contains hazardous material which requires special freight handling. Additional charges may apply.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.