Learn More
Ace Glass, Inc. 125mL separatory funnel with locking stopper
SDP
Catalog No. 50974699
Encompass_Preferred
125ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-699 Supplier Ace Glass, Inc. Supplier No. 723157
125ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.