Learn More
Waters Corp Waters Corporation Magnesium Cation Calibration Standard, 1000 mg/L, Unpres., 125 mL
Catalog No. 50466722 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Magnesium Cation Calibration Standard, with a concentration of 1000 mg/L in an H2O matrix, is designed for cation analysis by Ion Chromatography. It can be used for primary calibration or to prepare second source calibration check standards, ensuring accurate and reliable results.
  • Used for cation analysis by Ion Chromatography.
  • Utilized for primary calibration.
  • Prepares second source calibration check standards.
  • Traceable to NIST Standard Reference Materials (where available).
  • Guaranteed stable for one year.
  • Undergoes strict QC processes for chemical purity and stability.
  • Liquid formulation.
  • Volume: 125 mL.
  • Supplied as 1 pk.
Catalog No. 50-466-722 Supplier Waters Corp Supplier No. K11
Add to Cart
Add to Cart
This Magnesium Cation Calibration Standard, with a concentration of 1000 mg/L in an H2O matrix, is designed for cation analysis by Ion Chromatography. It can be used for primary calibration or to prepare second source calibration check standards, ensuring accurate and reliable results.
  • Used for cation analysis by Ion Chromatography.
  • Utilized for primary calibration.
  • Prepares second source calibration check standards.
  • Traceable to NIST Standard Reference Materials (where available).
  • Guaranteed stable for one year.
  • Undergoes strict QC processes for chemical purity and stability.
  • Liquid formulation.
  • Volume: 125 mL.
  • Supplied as 1 pk.