Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Trace Metal Standard, Mercury, 1000 mg/L, 125 mL

Catalog No. 50465850 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Mercury QC Trace Metal Standard is provided at a concentration of 1000 mg/L in an HNO3 matrix. It is suitable for use with ICP-OES, ICP-MS, and AA methods, and one 15 mL screw-cap vial can be diluted to yield up to 2 L.

  • Concentration: 1000 mg/L
  • Volume: 125 mL
  • Suitable for ICP-OES, ICP-MS, and AA methods
  • One 15 mL screw-cap vial yields up to 2 L after dilution
  • Components: Mercury
  • Molecular formula: Hg

Catalog No. 50-465-850 Supplier Waters Corp Supplier No. 027
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This Mercury QC Trace Metal Standard is provided at a concentration of 1000 mg/L in an HNO3 matrix. It is suitable for use with ICP-OES, ICP-MS, and AA methods, and one 15 mL screw-cap vial can be diluted to yield up to 2 L.

  • Concentration: 1000 mg/L
  • Volume: 125 mL
  • Suitable for ICP-OES, ICP-MS, and AA methods
  • One 15 mL screw-cap vial yields up to 2 L after dilution
  • Components: Mercury
  • Molecular formula: Hg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.