Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Trace Metal Standard, Nickel, 1000 mg/L, 125 mL

Catalog No. 50465852 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product is a Nickel, QC Trace Metal Standard with a concentration of 1000 mg/L in an HNO3 matrix. It is designed for use with ICP-OES, ICP-MS, and AA methods. This liquid standard is intended for environmental applications.

  • Concentration of 1000 mg/L.
  • Volume of 125 mL.
  • Formulated as a liquid.
  • Intended for environmental applications.
  • Compatible with ICP-OES, ICP-MS, and AA methods.
  • One 15 mL screw-cap vial yields up to 2 liters after dilution, offering extended use.
  • Contains hazardous material, requiring special freight handling.

Catalog No. 50-465-852 Supplier Waters Corp Supplier No. 029
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This product is a Nickel, QC Trace Metal Standard with a concentration of 1000 mg/L in an HNO3 matrix. It is designed for use with ICP-OES, ICP-MS, and AA methods. This liquid standard is intended for environmental applications.

  • Concentration of 1000 mg/L.
  • Volume of 125 mL.
  • Formulated as a liquid.
  • Intended for environmental applications.
  • Compatible with ICP-OES, ICP-MS, and AA methods.
  • One 15 mL screw-cap vial yields up to 2 liters after dilution, offering extended use.
  • Contains hazardous material, requiring special freight handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.