Learn More
Waters Corp Ion Calibration Standard, Nitrite as N, 1000 mg/L, 125 mL
Catalog No. 50465868 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ion calibration standard is designed for primary calibration or to prepare second source calibration check standards.
  • Nitrite as N
  • 1000 mg/L concentration
  • 125 mL volume
  • H2O matrix
  • Traceable to NIST Standard Reference Materials
  • Guaranteed stable for one year
Catalog No. 50-465-868 Supplier Waters Corp Supplier No. 053
This ion calibration standard is designed for primary calibration or to prepare second source calibration check standards.
  • Nitrite as N
  • 1000 mg/L concentration
  • 125 mL volume
  • H2O matrix
  • Traceable to NIST Standard Reference Materials
  • Guaranteed stable for one year