Learn More
Ace Glass, Inc. 125mL separatory funnel body only, pear shaped, #11Ace-Thred bottom, includes 2mm bore PTFE stopcock plug, 7223-40 #11 PTFE drip tip & 12632-16 size 16 PTFE stopper not included
SDP
Catalog No. 50974663
Encompass_Preferred
125ml pear-shaped, separatory funnel body. Funnel has a 2mm bore PTFE stopcock plug and a #11 Ace-Thred bottom which creates a leak tight connection without the need for an o-ring.
Catalog No. 50-974-663 Supplier Ace Glass, Inc. Supplier No. 722307
125ml pear-shaped, separatory funnel body. Funnel has a 2mm bore PTFE stopcock plug and a #11 Ace-Thred bottom which creates a leak tight connection without the need for an o-ring.