Learn More
Ace Glass, Inc. 125mL separatory funnel, pear shaped, complete with #11removable PTFE drip tip and 2mm bore PTFE stopcock plug, 12632-16 size 16 PTFE stopper not included
SDP
Catalog No. 50979211
Encompass_Preferred
125ml pear-shaped, separatory funnel complete with #11 removable PTFE drip tip and 2mm bore PTFE stopcock plug. Drip tip is detachable, difficult to break and easy to clean.
Catalog No. 50-979-211 Supplier Ace Glass, Inc. Supplier No. 722350
125ml pear-shaped, separatory funnel complete with #11 removable PTFE drip tip and 2mm bore PTFE stopcock plug. Drip tip is detachable, difficult to break and easy to clean.