Learn More
Triforest Enterprises 125ML autoclaveable polycarbonate square media bottle
Catalog No. NC2401706
Encompass_Preferred
TriForest Polycarbonate labware is a safer alternative to glass. It is ideal for steam sterilization of all biologicals, liquids, reagents, media, buffers, and aqueous solutions. Manufactured in a Class 10K cleanroom in an ISO 9001 facility with virgin Polycarbonate resin that is durable, chemically inert, non-leaching, and optically superior. Our square media bottles withstand repeated autoclaving, and are optimal for all microbial culture media production and preparation of terminally sterile fluidsblue autoclavable caps.
Catalog No. NC2401706 Supplier Triforest Enterprises Supplier No. BPC0125
TriForest Polycarbonate labware is a safer alternative to glass. It is ideal for steam sterilization of all biologicals, liquids, reagents, media, buffers, and aqueous solutions. Manufactured in a Class 10K cleanroom in an ISO 9001 facility with virgin Polycarbonate resin that is durable, chemically inert, non-leaching, and optically superior. Our square media bottles withstand repeated autoclaving, and are optimal for all microbial culture media production and preparation of terminally sterile fluidsblue autoclavable caps.