Learn More
Waters Corp Waters Ion Calibration Standard, Perchlorate, 1000 mg/L, 125 mL
Catalog No. 50465876 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This perchlorate ion calibration standard, at 1000 mg/L, is designed for use as a primary calibration standard or to prepare second source calibration check standards. It is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.
  • Ion calibration standard
  • Can be used for primary calibration or to prepare second source calibration check standards
  • Traceable to NIST Standard Reference Materials (where available)
  • Guaranteed stable for one year
Catalog No. 50-465-876 Supplier Waters Corp Supplier No. 068
This perchlorate ion calibration standard, at 1000 mg/L, is designed for use as a primary calibration standard or to prepare second source calibration check standards. It is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.
  • Ion calibration standard
  • Can be used for primary calibration or to prepare second source calibration check standards
  • Traceable to NIST Standard Reference Materials (where available)
  • Guaranteed stable for one year