Learn More
Bellco Glass, Inc. 125ml PETG Erlenmeyer Flask, Flat base, Vent Cap, Sterile,1/pk, 24/cs
SDP
Catalog No. 502329527
Encompass_Preferred
125ml PETG Erlenmeyer Flask, Flat base, Vent Cap, Sterile,1/pk, 24/cs. Ideal for shaker culture applications. There are two kinds of bottle materials, both PETG and PC are available. PC bottle or PEFG bottleSterilized by E-BeamNonpyrogenicIndividually wrapped in a plastic bag.Both vent caps and plug seal caps are available.Vent caps with 0.22um hydrophobic filters for gas exchange without contamination.
Catalog No. 50-232-9527 Supplier Bellco Glass, Inc. Supplier No. 451400182
125ml PETG Erlenmeyer Flask, Flat base, Vent Cap, Sterile,1/pk, 24/cs. Ideal for shaker culture applications. There are two kinds of bottle materials, both PETG and PC are available. PC bottle or PEFG bottleSterilized by E-BeamNonpyrogenicIndividually wrapped in a plastic bag.Both vent caps and plug seal caps are available.Vent caps with 0.22um hydrophobic filters for gas exchange without contamination.