Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Phosphorus Standard, 1000 mg/L, 125 mL, H2O

Catalog No. 50465871 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Phosphorus Ion Calibration Standard, 1000 mg/L in an H2O matrix, can be used for primary calibration or for preparing second source calibration check standards. It offers guaranteed stability for one year and is traceable to NIST Standard Reference Materials, wherever such materials are available.

  • Liquid formulation
  • Concentration: 1000 mg/L
  • Volume: 125 mL
  • H2O matrix (Ion Calibration Standard)
  • Guaranteed stable for one year
  • Traceable to NIST Standard Reference Materials, where available
  • Application: Environment
  • Product type: Standards

Catalog No. 50-465-871 Supplier Waters Corp Supplier No. 063
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Phosphorus Ion Calibration Standard, 1000 mg/L in an H2O matrix, can be used for primary calibration or for preparing second source calibration check standards. It offers guaranteed stability for one year and is traceable to NIST Standard Reference Materials, wherever such materials are available.

  • Liquid formulation
  • Concentration: 1000 mg/L
  • Volume: 125 mL
  • H2O matrix (Ion Calibration Standard)
  • Guaranteed stable for one year
  • Traceable to NIST Standard Reference Materials, where available
  • Application: Environment
  • Product type: Standards

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.