Learn More
Ace Glass, Inc. 125mL gas wash stopper only, porosity A
SDP
Catalog No. 50979107
Encompass_Preferred
125mL gas wash stopper with porosity A (145-174micron) fritted disc.
Catalog No. 50-979-107 Supplier Ace Glass, Inc. Supplier No. 716260
125mL gas wash stopper with porosity A (145-174micron) fritted disc.