Learn More
Ace Glass, Inc. 125mL complete gas washing bottle, porosity B
SDP
Supplier:  Ace Glass, Inc. 716304
Encompass_Preferred
125mL gas washing bottle with porosity B (70-100 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D
Catalog No. 50-974-110