Learn More
Ace Glass, Inc. 125mL gas wash stopper only, porosity B
SDP
Catalog No. 50974103
Encompass_Preferred
125mL gas wash stopper with porosity B (70-100micron) fritted disc.
Catalog No. 50-974-103 Supplier Ace Glass, Inc. Supplier No. 716262
125mL gas wash stopper with porosity B (70-100micron) fritted disc.