Learn More
Ace Glass, Inc. 125mL complete gas washing bottle, porosity C
SDP
Catalog No. 50974111
Encompass_Preferred
125mL gas washing bottle with porosity C (25-50 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.
Catalog No. 50-974-111 Supplier Ace Glass, Inc. Supplier No. 716306
125mL gas washing bottle with porosity C (25-50 micron) 25mm fritted disc diameter, and 40/35 joint. Inlet/outlet arms are 8mm O.D.