Learn More
CELLTREAT Scientific Products 125mL Wide Mouth Bottle, Round, PP, Non-sterile, 48/CS
SDP
Catalog No. 501535081
Encompass_Preferred
CELLTREAT Scientific Products 125mL Wide Mouth Bottle. Round shape and easy-pour wide mouth design. Bottle and cap manufactured from virgin polypropylene for excellent chemical resistance with durable, rigid walls. Features liner-less leak proof screw top closure. Bottle and cap come fully assembled. Ideal for specimen and chemical storage, storage of laboratory reagents and biological buffers and for sample shipment and packaging. Non-sterile; autoclavable. Every case is printed with the lot number for quality traceability. Individually packed in bags, 24/case.
Catalog No. 50-153-5081 Supplier CELLTREAT Scientific Products Supplier No. 229795
CELLTREAT Scientific Products 125mL Wide Mouth Bottle. Round shape and easy-pour wide mouth design. Bottle and cap manufactured from virgin polypropylene for excellent chemical resistance with durable, rigid walls. Features liner-less leak proof screw top closure. Bottle and cap come fully assembled. Ideal for specimen and chemical storage, storage of laboratory reagents and biological buffers and for sample shipment and packaging. Non-sterile; autoclavable. Every case is printed with the lot number for quality traceability. Individually packed in bags, 24/case.