Learn More
Ace Glass, Inc. 125mL graduated pressure equalizing funnel, 24/40 top, glass plug
SDP
Supplier:  Ace Glass, Inc. 729650
Encompass_Preferred
125mL graduated pressure equalizing funnel featuring a 24/40 standard taper joint at top. Cylindrical funnel features standard taper joints at the top and bottom, pressuring equalizing arm and double scale graduations calibrated from stopcock. Also featured is a drip system that does not extend beyond the joint to eliminate possible sources of breakage. Glass plug included.
Catalog No. 50-974-771